siRNA Details
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 4 | 1434 | ||
GenBank mRNA | AK082374.1 | - | 4 | 1436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
