siRNA Details
Gene Symbol: 1810032O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037979.1 | - | 5 | 910 | ||
BC055707.1 | - | 5 | 904 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_025472.1 | - | 910 | Chr. - - [-] on Build |
