siRNA Details
Gene Symbol: Dtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029545.2 | NP_083821.1 | 3 | 772 | ||
GenBank mRNA | AK018311.1 | - | 3 | 771 | ||
AK032228.1 | - | 3 | 770 | |||
AK042771.1 | - | 3 | 773 | |||
BC089589.1 | - | 3 | 772 | 6760937 |
Gene Symbol: 5830417I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031583.1 | - | 1 | 2393 | ||
AK049055.1 | - | 3 | 3816 |
Gene Symbol: Anapc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025514.2 | NP_079790.1 | 4 | 727 | ||
GenBank mRNA | AK009172.1 | - | 4 | 727 | ||
AK012280.1 | - | 4 | 595 | |||
AK088376.1 | - | 4 | 675 |
Gene Symbol: Gm19897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040398.1 | - | 4 | 1529 |
Gene Symbol: Ints6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008715.2 | NP_032741.2 | 19 | 4497 | ||
GenBank mRNA | BC059263.1 | - | 19 | 4497 | 6409580 |
Gene Symbol: Mettl5os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016085.1 | - | 1 | 352 | ||
AK050748.1 | - | 1 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
