siRNA Details
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 6 | 647 | ||
NM_001258332.1 | NP_001245261.1 | 4 | 522 | |||
GenBank mRNA | AK096026.1 | - | 3 | 306 | ||
AK096137.1 | - | 5 | 686 | |||
AK293212.1 | - | 4 | 805 | |||
AK300070.1 | - | 6 | 536 | |||
AK303279.1 | - | 4 | 475 | |||
AK310049.1 | - | 3 | 880 | |||
AK314340.1 | - | 6 | 629 | |||
BC015045.2 | - | 6 | 572 | 3922902 | ||
BI910081.1 | - | 6 | 549 | |||
BT009852.1 | - | 6 | 531 | |||
KU178089.1 | - | 6 | 531 | |||
KU178090.1 | - | 6 | 598 | |||
M60091.1 | - | 6 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
