siRNA Details
Gene Symbol: Teddm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025634.3 | NP_079910.1 | 1 | 281 | ||
GenBank mRNA | AK009367.1 | - | 1 | 225 | ||
AK009496.1 | - | 1 | 239 | |||
AK009566.1 | - | 1 | 283 | |||
AK009635.1 | - | 1 | 243 | |||
AK009751.1 | - | 1 | 341 | |||
AK009766.1 | - | 1 | 233 | |||
AK009777.1 | - | 1 | 225 | |||
AK009984.1 | - | 1 | 231 | |||
AK009997.1 | - | 1 | 280 | |||
AK010016.1 | - | 1 | 242 | |||
BC115655.1 | - | 1 | 232 | 40103183 | ||
BC115656.1 | - | 1 | 231 | 40103184 |
