siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 4 | 405 | ||
NM_001025445.2 | NP_001020616.1 | 4 | 400 | |||
NM_025545.4 | NP_079821.3 | 4 | 540 | |||
GenBank mRNA | AK005286.1 | - | 3 | 357 | ||
AK010516.1 | - | 4 | 387 | |||
AK017522.1 | - | 3 | 370 | |||
AK077351.1 | - | 4 | 541 | |||
AY040780.1 | - | 4 | 551 | |||
AY208844.1 | - | 4 | 552 | |||
BB860888.1 | - | 4 | 408 | |||
BC021872.1 | - | 4 | 398 | 3994375 | ||
BC068309.1 | - | 3 | 313 | |||
BC071213.1 | - | 4 | 392 |
