siRNA Details
Gene Symbol: AGXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000030.2 | NP_000021.1 | 2 | 312 | ||
GenBank mRNA | AB292648.1 | - | 2 | 213 | ||
AF348451.1 | - | 2 | 103 | |||
AK292754.1 | - | 2 | 233 | |||
AK298400.1 | - | 2 | 98 | |||
AK298420.1 | - | 2 | 228 | |||
AK315331.1 | - | 2 | 260 | |||
AK316495.1 | - | 2 | 98 | |||
BC132819.1 | - | 2 | 309 | 40146841 | ||
D13368.1 | - | 2 | 299 | |||
X53414.1 | - | 2 | 313 | |||
X56092.1 | - | 2 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
