siRNA Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025590.4 | NP_079866.2 | 5, 6 | 600 | ||
GenBank mRNA | AF416923.1 | - | 5, 6 | 538 | ||
AK031588.1 | - | 5, 6 | 517 | |||
AK038546.1 | - | 5, 6 | 538 | |||
AK048670.1 | - | 5, 6 | 600 | |||
AK052332.1 | - | 5, 6 | 616 | |||
AK122350.1 | - | 4, 5 | 387 | |||
BC042492.1 | - | 5, 6 | 535 | 4240966 |
