siRNA Details
Gene Symbol: ALAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000032.4 | NP_000023.2 | 2 | 308 | ||
NM_001037967.3 | NP_001033056.1 | 2 | 308 | |||
NM_001037968.3 | NP_001033057.1 | 3 | 415 | |||
XM_005261995.3 | XP_005262052.1 | 3 | 328 | |||
XM_011530771.1 | XP_011529073.1 | 2 | 221 | |||
XM_017029354.1 | XP_016884843.1 | 3 | 329 | |||
GenBank mRNA | AK290565.1 | - | 1 | 181 | ||
AK291589.1 | - | 2 | 221 | |||
AK313118.1 | - | 2 | 308 | |||
BC030230.2 | - | 2 | 294 | 5226194 | ||
BP284609.1 | - | 2 | 308 | |||
X56352.1 | - | 1 | 167 | |||
X60364.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
