siRNA Details
Gene Symbol: Fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025626.4 | NP_079902.1 | 3 | 450 | ||
GenBank mRNA | AK013932.2 | - | 3 | 415 | ||
AK019815.1 | - | 3 | 362 | |||
AK032875.1 | - | 3 | 451 | |||
AK168759.1 | - | 3 | 416 | |||
BC021353.1 | - | 3 | 416 | 5006672 |
