siRNA Details
Gene Symbol: 3110001I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025653.2 | NP_079929.1 | 2 | 397 | ||
GenBank mRNA | AK012939.1 | - | 2 | 398 | ||
AK013950.1 | - | 2 | 468 | |||
AK031567.1 | - | 2 | 431 | |||
BC038880.1 | - | 1 | 216 | 1477704 |
