siRNA Details
Gene Symbol: Ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025671.2 | NP_079947.1 | 2 | 170 | ||
GenBank mRNA | AK004916.1 | - | 2 | 169 | ||
AK004943.1 | - | 2 | 171 | |||
AK017497.1 | - | 2 | 171 | |||
AK018534.1 | - | 2 | 171 | |||
AK168946.1 | - | 2 | 179 | |||
BC022762.1 | - | 1 | 18 | |||
BC069872.1 | - | 2 | 173 | 4219293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
