siRNA Details
Gene Symbol: Mcm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291054.1 | NP_001277983.1 | 2 | 442 | ||
NM_025676.4 | NP_079952.2 | 2 | 442 | |||
GenBank mRNA | AK012845.1 | - | 2 | 401 | ||
AK017607.1 | - | 2 | 340 | |||
AK019268.1 | - | 2 | 401 | |||
AK133858.1 | - | 2 | 399 | |||
BC046780.1 | - | 2 | 389 | 5716136 | ||
BC052070.1 | - | 2 | 248 | 5716125 |
