siRNA Details
Gene Symbol: Spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025697.3 | NP_079973.1 | 2 | 425 | ||
GenBank mRNA | AK018044.1 | - | 2 | 303 | ||
AK018153.1 | - | 2 | 426 | |||
AK018290.1 | - | 2 | 275 | |||
AK137685.1 | - | 274 | ||||
AK150789.1 | - | 2 | 300 | |||
AK167306.1 | - | 2 | 267 | |||
AK169059.1 | - | 2 | 274 | |||
BC037015.1 | - | 2 | 259 | 5344409 |
