siRNA Details
Gene Symbol: 4933415F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025746.2 | NP_080022.2 | 1 | 306 | ||
GenBank mRNA | AK016819.1 | - | 1 | 307 | ||
BC115556.1 | - | 1 | 307 | 40101816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
