siRNA Details
Gene Symbol: Gm9758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198666.3 | NP_941068.1 | 2 | 266 | ||
GenBank mRNA | BC048454.1 | - | 2 | 266 | 6741646 |
Gene Symbol: 4930572O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076955.1 | - | 2 | 250 |
Gene Symbol: Gm10354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281514.1 | NP_001268443.1 | 2 | 230 | ||
GenBank mRNA | CJ042498.1 | - | 231 |
Gene Symbol: Gm17019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182957.1 | NP_892002.1 | 2 | 275 | ||
GenBank mRNA | CJ047174.1 | - | 231 |
Gene Symbol: Speer4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281511.1 | NP_001268440.1 | 2 | 275 | ||
GenBank mRNA | AK006011.1 | - | 276 |
Gene Symbol: Speer4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025759.3 | NP_080035.2 | 2 | 240 | ||
GenBank mRNA | AK017002.1 | - | 2 | 241 |
Gene Symbol: Speer4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122661.1 | NP_001116133.1 | 2 | 275 | ||
GenBank mRNA | BC100449.1 | - | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
