siRNA Details
Gene Symbol: 4931417E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025737.3 | NP_080013.1 | 1 | 352 | ||
GenBank mRNA | AK005631.1 | - | 1 | 277 | ||
AK016460.1 | - | 1 | 353 | |||
BC048456.1 | - | 1 | 304 | 6771342 |
