siRNA Details
Gene Symbol: Speer4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281511.1 | NP_001268440.1 | 3 | 503 |
Gene Symbol: 4930572O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076955.1 | - | 3 | 478 |
Gene Symbol: Gm17019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182957.1 | NP_892002.1 | 3 | 503 |
Gene Symbol: Gm9758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198666.3 | NP_941068.1 | 3 | 494 | ||
GenBank mRNA | BC048454.1 | - | 3 | 494 | 6741646 |
Gene Symbol: Speer4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025759.3 | NP_080035.2 | 3 | 468 | ||
GenBank mRNA | AK017002.1 | - | 3 | 469 |
Gene Symbol: Speer4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122661.1 | NP_001116133.1 | 3 | 503 | ||
GenBank mRNA | BC100449.1 | - | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
