siRNA Details
Gene Symbol: PRKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282480.1 | NP_001269409.1 | 2 | 110 | ||
NM_001282481.1 | NP_001269410.1 | 2 | 110 | |||
NM_001282482.1 | NP_001269411.1 | 2 | 110 | |||
NM_001282483.1 | NP_001269412.1 | 2 | 110 | |||
NM_001282485.1 | NP_001269414.1 | 7 | 1161 | |||
NM_006259.2 | NP_006250.1 | 7 | 1161 | |||
XM_005263126.3 | XP_005263183.1 | 7 | 1596 | |||
XM_017008413.1 | XP_016863902.1 | 7 | 1598 | |||
XM_017008414.1 | XP_016863903.1 | 7 | 1593 | |||
XM_017008415.1 | XP_016863904.1 | 7 | 1003 | |||
XM_017008416.1 | XP_016863905.1 | 7 | 1515 | |||
XM_017008417.1 | XP_016863906.1 | 6 | 979 | |||
XM_017008418.1 | XP_016863907.1 | 6 | 979 | |||
GenBank mRNA | AK296979.1 | - | 2 | 97 | ||
AK297673.1 | - | 7 | 1108 | |||
AK316140.1 | - | 2 | 110 | |||
D70899.1 | - | 6 | 966 | |||
DA462272.1 | - | 2 | 110 | |||
DC348270.1 | - | 2 | 98 | |||
X94612.1 | - | 6 | 980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
