siRNA Details
Gene Symbol: Rpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025683.2 | NP_079959.2 | 6 | 2395 | ||
GenBank mRNA | AK017774.1 | - | 6 | 2396 | ||
AK029088.1 | - | 6 | 2394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
