siRNA Details
Gene Symbol: Col27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025685.3 | NP_079961.3 | 61 | 6371 | ||
GenBank mRNA | AK017766.1 | - | 2 | 1073 | ||
AK030038.1 | - | 39 | 4258 | |||
AK173283.1 | - | 37 | 2995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
