siRNA Details
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025693.4 | NP_079969.1 | 5 | 1016 | ||
GenBank mRNA | AK007685.1 | - | 5 | 1011 | ||
AK013518.1 | - | 4 | 841 | |||
AK160292.1 | - | 5 | 937 | |||
AK214562.1 | - | 1 | 27 | |||
BC011512.1 | - | 5 | 997 | |||
BC019770.1 | - | 5 | 971 | 5124769 |
