siRNA Details
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277340.1 | NP_001264269.1 | 1 | 1438 | ||
GenBank mRNA | AK006425.1 | - | 1 | 1425 | ||
AK016920.1 | - | 1 | 1405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
