siRNA Details
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025800.3 | NP_080076.1 | 1, 2 | 432 | ||
GenBank mRNA | AK002671.1 | - | 1, 2 | 379 | ||
AK015709.1 | - | 205 | ||||
AK028603.1 | - | 1, 2 | 384 | |||
AK032462.1 | - | 1, 2 | 383 | |||
AK053657.1 | - | 1, 2 | 333 | |||
AK077194.1 | - | 1, 2 | 257 | |||
AK150281.1 | - | 1, 2 | 435 | |||
BC069885.1 | - | 1, 2 | 405 | 3671082 | ||
BC069886.1 | - | 1, 2 | 377 | 6310133 |
Gene Symbol: Ppp1r2-ps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100445.1 | - | 1 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
