siRNA Details
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025815.2 | NP_080091.1 | 7 | 549 | ||
GenBank mRNA | AK004559.1 | - | 7 | 537 | ||
AK010227.1 | - | 7 | 549 | |||
AK013407.1 | - | 6 | 457 | |||
BC017626.1 | - | 6 | 443 | 4502073 | ||
BC138745.1 | - | 7 | 548 | 8861767 | ||
BC138746.1 | - | 7 | 548 | 8861768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
