siRNA Details
Gene Symbol: TAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000116.4 | NP_000107.1 | 4, 5 | 704 | ||
NM_001303465.1 | NP_001290394.1 | 4, 5 | 758 | |||
NM_181312.3 | NP_851829.1 | 4, 5 | 704 | |||
XM_006724836.1 | XP_006724899.1 | 4, 5 | 735 | |||
XM_006724841.3 | XP_006724904.1 | 4, 5 | 456 | |||
XM_011531191.2 | XP_011529493.1 | 3, 4 | 894 | |||
XM_017029764.1 | XP_016885253.1 | 2, 3 | 233 | |||
GenBank mRNA | AK291848.1 | - | 4, 5 | 572 | ||
AY231462.1 | - | 4, 5 | 386 | |||
AY231464.1 | - | 4, 5 | 386 | |||
AY258036.1 | - | 4, 5 | 386 | |||
AY258038.1 | - | 4, 5 | 386 | |||
CA488175.1 | - | 3, 4 | 300 | |||
DQ884408.1 | - | 4, 5 | 432 | |||
KU178349.1 | - | 4, 5 | 378 | |||
KU178350.1 | - | 4, 5 | 378 | |||
X92762.1 | - | 4, 5 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
