siRNA Details
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025852.3 | NP_080128.2 | 5 | 2453 | ||
GenBank mRNA | AK006228.1 | - | 1 | 446 | ||
AK007009.1 | - | 1 | 112 | |||
AK173108.1 | - | 5 | 2418 | |||
BC049901.1 | - | 2 | 256 | |||
BC052424.1 | - | 5 | 2435 | 6835200 | ||
BC054843.1 | - | 5 | 2411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
