siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293644.1 | NP_001280573.1 | 6 | 850 | ||
NM_025779.3 | NP_080055.2 | 7 | 1011 | |||
GenBank mRNA | AK013032.1 | - | 3 | 319 | ||
AK018492.1 | - | 7 | 888 | |||
AK150724.1 | - | 7 | 909 | |||
AK150995.1 | - | 7 | 885 | |||
AK151201.1 | - | 7 | 909 | |||
AK151915.1 | - | 7 | 922 | |||
AK152326.1 | - | 6 | 721 | |||
AK154338.1 | - | 7 | 904 | |||
BC049571.1 | - | 7 | 1011 | 6588325 | ||
BX515874.1 | - | 2 | 62 |
