siRNA Details
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025852.3 | NP_080128.2 | 6 | 2605 | ||
GenBank mRNA | AK006228.1 | - | 2 | 598 | ||
AK007009.1 | - | 2 | 264 | |||
AK079120.1 | - | 2 | 119 | |||
AK173108.1 | - | 6 | 2570 | |||
BC049901.1 | - | 3 | 408 | |||
BC052424.1 | - | 6 | 2587 | 6835200 | ||
BC054843.1 | - | 6 | 2563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
