siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293644.1 | NP_001280573.1 | 7 | 1081 | ||
NM_025779.3 | NP_080055.2 | 8 | 1242 | |||
GenBank mRNA | AK013032.1 | - | 4 | 550 | ||
AK018492.1 | - | 8 | 1119 | |||
AK150724.1 | - | 8 | 1140 | |||
AK150995.1 | - | 8 | 1116 | |||
AK151201.1 | - | 8 | 1140 | |||
AK151915.1 | - | 8 | 1153 | |||
AK152326.1 | - | 7 | 952 | |||
AK154338.1 | - | 8 | 1135 | |||
BC049571.1 | - | 8 | 1242 | 6588325 | ||
BX515874.1 | - | 3 | 293 |
