siRNA Details
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081274.1 | NP_001074743.1 | 7 | 721 | ||
GenBank mRNA | AK002894.1 | - | 7 | 683 | ||
AK036820.1 | - | 1 | 1993 | |||
AK145602.1 | - | 7 | 685 | |||
AK150210.1 | - | 7 | 684 | |||
AK150490.1 | - | 7 | 680 | |||
AK151380.1 | - | 7 | 680 | |||
AK151993.1 | - | 7 | 680 | |||
AK152758.1 | - | 7 | 680 | |||
AK152894.1 | - | 7 | 680 | |||
AK153409.1 | - | 7 | 677 | |||
AK155027.1 | - | 7 | 680 | |||
AK161869.1 | - | 6 | 2642 | |||
AK166733.1 | - | 7 | 675 | |||
AK166947.1 | - | 7 | 657 | |||
AK167215.1 | - | 7 | 675 | |||
AK168251.1 | - | 7 | 680 | |||
AK216999.1 | - | 1 | 74 | |||
BC008646.1 | - | 7 | 652 | 3155147 | ||
BC011329.1 | - | 7 | 670 | 3984654 | ||
BC014793.1 | - | 7 | 689 | 3155100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
