siRNA Details
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025815.2 | NP_080091.1 | 20 | 2664 | ||
GenBank mRNA | AK004559.1 | - | 20 | 2652 | ||
BC017626.1 | - | 19 | 2558 | 4502073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
