siRNA Details
Gene Symbol: FECH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000140.3 | NP_000131.2 | 2, 3 | 319 | ||
XM_011525882.2 | XP_011524184.1 | 2, 3 | 302 | |||
XM_017025614.1 | XP_016881103.1 | 2, 3 | 290 | |||
GenBank mRNA | AK223190.1 | - | 2, 3 | 194 | ||
AK292937.1 | - | 2, 3 | 203 | |||
AK299643.1 | - | 2, 3 | 228 | |||
BC039841.1 | - | 2, 3 | 286 | 6042757 | ||
BG772545.1 | - | 2, 3 | 355 | |||
BT019958.1 | - | 2, 3 | 202 | |||
BT019959.1 | - | 2, 3 | 202 | |||
BX571744.1 | - | 2, 3 | 199 | |||
D00726.1 | - | 2, 3 | 231 | |||
DA990386.1 | - | 2, 3 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
