siRNA Details
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025852.3 | NP_080128.2 | 6, 7 | 2650 | ||
GenBank mRNA | AK006228.1 | - | 2, 3 | 643 | ||
AK007009.1 | - | 2, 3 | 309 | |||
AK079120.1 | - | 2, 3 | 164 | |||
AK173108.1 | - | 6, 7 | 2615 | |||
BC049901.1 | - | 3, 4 | 453 | |||
BC052424.1 | - | 6, 7 | 2632 | 6835200 | ||
BC054843.1 | - | 6, 7 | 2608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
