siRNA Details
Gene Symbol: Tmem134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078649.1 | NP_001072117.1 | 1 | 209 | ||
NM_025889.2 | NP_080165.1 | 1 | 209 | |||
GenBank mRNA | AK002220.1 | - | 1 | 203 | ||
AK008515.1 | - | 1 | 182 | |||
AK009263.1 | - | 1 | 211 | |||
AK009745.1 | - | 1 | 185 | |||
AK010323.1 | - | 1 | 211 | |||
AK129014.1 | - | 1 | 182 | |||
AK166519.1 | - | 1 | 212 | |||
BC026765.1 | - | 1 | 191 | 4166287 | ||
CF983315.1 | - | 1 | 235 |
