siRNA Details
Gene Symbol: Tex40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039494.2 | NP_001034583.1 | 2 | 238 | ||
GenBank mRNA | AK005644.1 | - | 2 | 217 | ||
AK006034.1 | - | 2 | 176 | |||
AK006134.1 | - | 2 | 241 | |||
AK007089.1 | - | 2 | 214 | |||
BC048452.1 | - | 2 | 175 | |||
BC061075.1 | - | 2 | 180 | |||
BC172038.1 | - | 1 | 395 | |||
BC172041.1 | - | 1 | 395 | |||
BC172042.1 | - | 1 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
