siRNA Details
Gene Symbol: 1700011H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025956.4 | NP_080232.3 | 2 | 230 | ||
GenBank mRNA | AK005866.1 | - | 2 | 197 | ||
AK007176.1 | - | 2 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
