siRNA Details
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025965.3 | NP_080241.3 | 1, 2 | 194 | ||
GenBank mRNA | AB009295.1 | - | 1, 2 | 171 | ||
AK010884.1 | - | 1, 2 | 140 | |||
AK031538.1 | - | 1, 2 | 174 | |||
AK160285.1 | - | 1, 2 | 157 | |||
AK166235.1 | - | 1, 2 | 168 | |||
AK167793.1 | - | 1, 2 | 140 | |||
AK216988.1 | - | 13 | ||||
BB619494.1 | - | 1, 2 | 213 | |||
BC011255.1 | - | 1, 2 | 129 | 4017600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
