siRNA Details
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 2, 3 | 293 | ||
GenBank mRNA | AK012645.1 | - | 2, 3 | 286 | ||
AK013457.1 | - | 2, 3 | 337 | |||
AK133715.1 | - | 2, 3 | 292 | |||
AK153850.1 | - | 2, 3 | 271 | |||
BC023046.1 | - | 2, 3 | 164 | |||
BC050185.1 | - | 2, 3 | 273 | |||
BC064777.1 | - | 2, 3 | 277 | |||
BC080825.1 | - | 2, 3 | 285 | |||
BC094292.1 | - | 2, 3 | 283 | |||
CF907970.1 | - | 2, 3 | 271 |

More Information
Other Species Targeted

Gene Ontology Categories:
