siRNA Details
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025901.3 | NP_080177.1 | 3 | 427 | ||
GenBank mRNA | AK005152.1 | - | 3 | 331 | ||
AK008637.1 | - | 3 | 299 | |||
AK010875.1 | - | 3 | 365 | |||
AK021353.1 | - | 3 | 362 | |||
AK031110.1 | - | 3 | 362 | |||
AK088206.1 | - | 3 | 327 | |||
AK161764.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
