siRNA Details
Gene Symbol: Zfp830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025884.4 | NP_080160.2 | 1 | 1010 | ||
GenBank mRNA | AK008475.1 | - | 1 | 1010 | ||
AK010353.1 | - | 1 | 1001 | |||
AK017640.1 | - | 1 | 990 | |||
AK050043.1 | - | 1 | 1005 | |||
AK132497.1 | - | 1 | 1011 | |||
AK148058.1 | - | 1 | 1011 | |||
AK163064.1 | - | 1 | 72 | |||
AK167656.1 | - | 1 | 1001 | |||
AK168417.1 | - | 1 | 990 | |||
BC024340.1 | - | 1 | 996 | 4981533 |
