siRNA Details
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 19 | 3333 | ||
GenBank mRNA | AK011184.1 | - | 7 | 1688 | ||
AK076553.1 | - | 5 | 1461 | |||
AK087936.1 | - | 7 | 1659 | |||
AK142132.1 | - | 13 | 2609 | |||
BC066150.1 | - | 8 | 1766 | |||
BC067400.1 | - | 8 | 1766 | |||
BC158104.1 | - | 18 | 3213 | 9007388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
