siRNA Details
Gene Symbol: Ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025942.2 | NP_080218.1 | 11 | 1284 | ||
GenBank mRNA | AK010817.1 | - | 11 | 1200 | ||
AK013055.1 | - | 11 | 1285 | |||
AK159873.1 | - | 11 | 1201 | |||
AK167458.1 | - | 11 | 1200 | |||
AK170982.1 | - | 11 | 1215 | |||
BC011318.1 | - | 11 | 1184 | 3985271 | ||
BC029207.1 | - | 11 | 1177 | 4009561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
