siRNA Details
Gene Symbol: D16Ertd472e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252438.1 | NP_001239367.1 | 4, 5 | 1221 | ||
NM_001252439.1 | NP_001239368.1 | 4, 5 | 988 | |||
NM_001252440.1 | NP_001239369.1 | 4, 5 | 953 | |||
NM_025967.4 | NP_080243.3 | 4, 5 | 1021 | |||
GenBank mRNA | AK009258.1 | - | 4, 5 | 1192 | ||
AK044670.1 | - | 4, 5 | 1022 | |||
BC019957.1 | - | 4, 5 | 912 | 3966397 |
