siRNA Details
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025572.1 | NP_001020743.1 | 3 | 372 | ||
GenBank mRNA | AK012645.1 | - | 3 | 365 | ||
AK013457.1 | - | 3 | 416 | |||
AK133715.1 | - | 3 | 371 | |||
AK153850.1 | - | 3 | 350 | |||
BC023046.1 | - | 3 | 243 | |||
BC050185.1 | - | 3 | 352 | |||
BC064777.1 | - | 3 | 356 | |||
BC080825.1 | - | 3 | 364 | |||
BC094292.1 | - | 3 | 362 | |||
CF907970.1 | - | 3 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
