siRNA Details
Gene Symbol: Nkain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025998.3 | NP_080274.2 | 5, 6 | 685 | ||
GenBank mRNA | AK011164.1 | - | 5, 6 | 818 | ||
AK011850.1 | - | 5, 6 | 616 | |||
AK076029.1 | - | 2, 3 | 173 | |||
AK184838.1 | - | 2, 3 | 138 | |||
AK187377.1 | - | 2, 3 | 138 | |||
AK187454.1 | - | 2, 3 | 138 | |||
AK205247.1 | - | 2, 3 | 138 | |||
AK205326.1 | - | 2, 3 | 138 | |||
AK205517.1 | - | 2, 3 | 137 | |||
AK206217.1 | - | 2, 3 | 138 | |||
AK206243.1 | - | 2, 3 | 138 | |||
AK208705.1 | - | 2, 3 | 138 | |||
AK208715.1 | - | 2, 3 | 138 | |||
AK208816.1 | - | 2, 3 | 137 | |||
AK209134.1 | - | 2, 3 | 138 | |||
AK209985.1 | - | 2, 3 | 138 | |||
AK210092.1 | - | 2, 3 | 138 | |||
AK210264.1 | - | 2, 3 | 138 | |||
AK211065.1 | - | 2, 3 | 137 | |||
AK211305.1 | - | 2, 3 | 138 | |||
AK211393.1 | - | 2, 3 | 138 | |||
AK211541.1 | - | 2, 3 | 138 | |||
AK211571.1 | - | 2, 3 | 142 | |||
AK213620.1 | - | 2, 3 | 138 | |||
AK215330.1 | - | 2, 3 | 138 | |||
AK216110.1 | - | 2, 3 | 138 | |||
AK216558.1 | - | 2, 3 | 138 | |||
AK216625.1 | - | 2, 3 | 138 | |||
AK216635.1 | - | 2, 3 | 138 | |||
AK217347.1 | - | 2, 3 | 138 | |||
AK218211.1 | - | 2, 3 | 138 | |||
AK218646.1 | - | 2, 3 | 138 | |||
AK218997.1 | - | 2, 3 | 138 | |||
AK219655.1 | - | 2, 3 | 139 | |||
AK220029.1 | - | 2, 3 | 139 | |||
BC116391.1 | - | 5, 6 | 615 | 40101865 | ||
BC116392.1 | - | 5, 6 | 632 | |||
EF058048.2 | - | 5, 6 | 738 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008764183.
Gene Ontology Categories:
