siRNA Details
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026036.3 | NP_080312.1 | 2 | 240 | ||
GenBank mRNA | AK012941.1 | - | 2 | 241 | ||
AK030885.1 | - | 2 | 220 | |||
AK128960.1 | - | 2 | 226 | |||
AK145743.1 | - | 2 | 221 | |||
AK150775.1 | - | 2 | 241 | |||
AK151092.1 | - | 2 | 241 | |||
AK151431.1 | - | 2 | 241 | |||
AK152448.1 | - | 2 | 242 | |||
AK152723.1 | - | 2 | 251 | |||
AK152817.1 | - | 2 | 251 | |||
AK153092.1 | - | 2 | 241 | |||
AK154481.1 | - | 2 | 227 | |||
AK159439.1 | - | 2 | 210 | |||
AK159735.1 | - | 2 | 235 | |||
AK165948.1 | - | 2 | 209 | |||
AK167499.1 | - | 2 | 227 | |||
AK167854.1 | - | 2 | 232 | |||
AK170905.1 | - | 2 | 242 | |||
AK171394.1 | - | 2 | 271 | |||
AY241868.1 | - | 2 | 277 | |||
BC027248.1 | - | 2 | 212 | 2811938 |
