siRNA Details
Gene Symbol: 3110070M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005613.1 | - | 1 | 446 | ||
AK014250.1 | - | 1 | 454 | |||
BC050963.1 | - | 1 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
