siRNA Details
Gene Symbol: 1700093K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110133.1 | NP_001103603.1 | 3 | 657 | ||
NM_026105.3 | NP_080381.2 | 3 | 339 | |||
GenBank mRNA | AK007052.1 | - | 3 | 245 | ||
AK007139.1 | - | 2 | 96 | |||
BC049567.1 | - | 3 | 339 | 6773879 | ||
CF578320.1 | - | 3 | 685 |
