siRNA Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286986.1 | NP_001273915.1 | 7, 8 | 861 | ||
NM_001286987.1 | NP_001273916.1 | 6, 7 | 739 | |||
NM_026013.3 | NP_080289.1 | 7, 8 | 864 | |||
GenBank mRNA | AK009940.1 | - | 7, 8 | 801 | ||
AK012044.1 | - | 7, 8 | 781 | |||
AK015888.1 | - | 7, 8 | 827 | |||
AK052824.1 | - | 6, 7 | 673 | |||
AK172455.1 | - | 7, 8 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
